
N-Phenethyl-4-piperidone-13C6 (NPP-13C6) solution

Empirical Formula (Hill Notation):
C713C6H17NO
Molecular Weight:
209.24
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing