Skip to Content
Merck
N,N-Dimethyl-4-nitrosoaniline ≥97%

N,N-Dimethyl-4-nitrosoaniline

N,N-Dimethyl-4-nitrosoaniline ≥97%
Synonym(s):
4-Nitroso-N,N-dimethylaniline
Linear Formula:
ONC6H4N(CH3)2
CAS Number:
Molecular Weight:
150.18
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing