![Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse clone MAPK-YT, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/313/841/a313d318-fa40-40f5-804c-0cda2bdd96cb/640/a313d318-fa40-40f5-804c-0cda2bdd96cb.jpg)
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse
![Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse clone MAPK-YT, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/313/841/a313d318-fa40-40f5-804c-0cda2bdd96cb/640/a313d318-fa40-40f5-804c-0cda2bdd96cb.jpg)
Synonym(s):
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing