![Monoclonal Anti-Human IgE antibody produced in mouse clone GE-1, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/195/841/6cff0f0b-6ad6-45d8-8dd5-b5ae3f75de0f/640/6cff0f0b-6ad6-45d8-8dd5-b5ae3f75de0f.jpg)
Monoclonal Anti-Human IgE antibody produced in mouse
![Monoclonal Anti-Human IgE antibody produced in mouse clone GE-1, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/195/841/6cff0f0b-6ad6-45d8-8dd5-b5ae3f75de0f/640/6cff0f0b-6ad6-45d8-8dd5-b5ae3f75de0f.jpg)
Synonym(s):
Monoclonal Anti-Human IgE
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing