Skip to Content
Merck
Mebeverine impurity standard British Pharmacopoeia (BP) Reference Standard

Mebeverine impurity standard

Mebeverine impurity standard British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Mebeverine hydrochloride
Empirical Formula (Hill Notation):
C25H35NO5 · HCl
CAS Number:
Molecular Weight:
466.01

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing