Skip to Content
Merck
L-Serine-13C3,15N 98 atom % 15N, 98 atom % 13C, 95% (CP)

L-Serine-13C3,15N

L-Serine-13C3,15N 98 atom % 15N, 98 atom % 13C, 95% (CP)
Synonym(s):
L-Serine-UL-13C,15N
Linear Formula:
HO13CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
109.06

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing