Skip to Content
Merck
L-Glutamine-13C5 98 atom % 13C, 95% (CP)

L-Glutamine-13C5

L-Glutamine-13C5 98 atom % 13C, 95% (CP)
Synonym(s):
(S)-2,5-Diamino-5-oxopentanoic acid-13C5, L-Glutamic acid 5-amide-13C5, L-Glutamine-ul-13C, 13C Labeled glutamine
Linear Formula:
H2N13CO(13CH2)213CH(NH2)13CO2H
CAS Number:
Molecular Weight:
151.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing