Skip to Content
Merck
KN-93 ≥98% (HPLC)

KN-93

KN-93 ≥98% (HPLC)
Synonym(s):
N-[2-[N-(4-Chlorocinnamyl)-N-methylaminomethyl]phenyl]-N-(2-hydroxyethyl)-4-methoxybenzenesulfonamide phosphate salt, N-[2-[[[3-(4′-Chlorophenyl)-2-propenyl]methylamino]methyl]phenyl]-N-(2-hydroxyethyl)-4′-methoxybenzenesulfonamide phosphate salt
Empirical Formula (Hill Notation):
C26H29ClN2O4S · H3PO4
CAS Number:
Molecular Weight:
599.03

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing