Skip to Content
Merck
Ketoprofen Related Compound D pharmaceutical secondary standard, certified reference material

Ketoprofen Related Compound D

Ketoprofen Related Compound D pharmaceutical secondary standard, certified reference material
Synonym(s):
1-(3-Benzoylphenyl)ethanone, 3-Acetylbenzophenone
Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25

Products

Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1