Skip to Content
Merck
Isotachysterol 3 solution 100 μg/mL in hexane, certified reference material, ampule of 1 mL, Cerilliant®

Isotachysterol 3 solution

Isotachysterol 3 solution 100 μg/mL in hexane, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
(S)-4-Methyl-3-((E)-2-((1R,7aR)-7a-Methyl-1-((R)-6-Methylheptan-2-yl)-2,3,5,6,7,7a-Hexahydro-1H-Inden-4-yl)vinyl)cyclohex-3-en-1-ol)
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64

Products

Brand
Formula Weight
Manufacturer
Format
Package Size