
HEPES-d18

Synonym(s):
4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid-d18
Empirical Formula (Hill Notation):
C8D18N2O4S
Molecular Weight:
256.42
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing