
Glutathione Impurity D

Synonym(s):
γ-Glu-Cys, γ-L-Glutamyl-L-cysteine, des-Gly-glutathione reduced form
Empirical Formula (Hill Notation):
C8H14N2O5S
CAS Number:
Molecular Weight:
250.27
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing