Skip to Content
MX
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gilding metal alloy, Cu85Zn15
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF25040674
Gilding metal alloy, Cu85Zn15,
100m wire, 0.711mm diameter, as drawn
Expand
View Pricing
GF03400322
Gilding metal alloy, Cu85Zn15,
50m wire, 0.711mm diameter, as drawn
Expand
View Pricing
GF12027204
Gilding metal alloy, Cu85Zn15,
20m wire, 0.711mm diameter, as drawn
Expand
View Pricing
GF52096215
Gilding metal alloy, Cu85Zn15,
10m wire, 0.711mm diameter, as drawn
Expand
View Pricing
GF40662536
Gilding metal alloy, Cu85Zn15,
1m wire, 0.711mm diameter, as drawn
Expand
View Pricing
GF31906199
Gilding metal alloy, Cu85Zn15,
10m wire, 0.061mm diameter, as drawn
Expand
View Pricing
GF88274458
Gilding metal alloy, Cu85Zn15,
20m wire, 0.061mm diameter, as drawn
Expand
View Pricing
GF76959253
Gilding metal alloy, Cu85Zn15,
100m wire, 0.061mm diameter, as drawn
Expand
View Pricing
GF82816768
Gilding metal alloy, Cu85Zn15,
5m wire, 0.711mm diameter, as drawn
Expand
View Pricing
GF97415679
Gilding metal alloy, Cu85Zn15,
50m wire, 0.061mm diameter, as drawn
Expand
View Pricing