Skip to Content
Merck
Eucalyptol (cineole) Pharmaceutical Secondary Standard; Certified Reference Material

Eucalyptol (cineole)

Eucalyptol (cineole) Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Eucalyptol, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1