Skip to Content
Merck
Ethyl 2-methyl-4-pentenoate ≥98%, stabilized, FG

Ethyl 2-methyl-4-pentenoate

Ethyl 2-methyl-4-pentenoate ≥98%, stabilized, FG
Linear Formula:
H2C=CHCH2CH(CH3)CO2C2H5
CAS Number:
Molecular Weight:
142.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing