Estriol impurity standard
Synonym(s):
Estriol, 1,3,5(10)-Estratriene-3,16α,17β-triol, 16α-Hydroxyestradiol, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene
Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing