Skip to Content
Merck
Eprosartan Related Compound B United States Pharmacopeia (USP) Reference Standard

Eprosartan Related Compound B

Eprosartan Related Compound B United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Thiophenepropionic acid, 3-(2-Thienyl)propionic acid
Empirical Formula (Hill Notation):
C7H8O2S
CAS Number:
Molecular Weight:
156.20

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1