
Dextropropoxyphene for system suitability

Synonym(s):
Dextropropoxyphene hydrochloride
Empirical Formula (Hill Notation):
C22H29NO2 · HCl
CAS Number:
Molecular Weight:
375.93
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing