
Desacetyl Diltiazem HCl

Synonym(s):
(d-cis-3-Hydroxy-2,3-dihydro-5-[2-dimethylamino)ethyl]-2-(p-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride
Empirical Formula (Hill Notation):
C20H24N2O3S · HCl
CAS Number:
Molecular Weight:
408.94
Products
Brand
Formula Weight
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing