![Copper phthalocyanine-3,4′,4″,4″′-tetrasulfonic acid tetrasodium salt Dye content 85 %](/deepweb/assets/sigmaaldrich/product/structures/140/401/c0c29e46-8b2d-49a8-83b3-3369f9936856/640/c0c29e46-8b2d-49a8-83b3-3369f9936856.png)
Copper phthalocyanine-3,4′,4″,4″′-tetrasulfonic acid tetrasodium salt
![Copper phthalocyanine-3,4′,4″,4″′-tetrasulfonic acid tetrasodium salt Dye content 85 %](/deepweb/assets/sigmaaldrich/product/structures/140/401/c0c29e46-8b2d-49a8-83b3-3369f9936856/640/c0c29e46-8b2d-49a8-83b3-3369f9936856.png)
Empirical Formula (Hill Notation):
C32H12CuN8O12S4 · 4Na
CAS Number:
Molecular Weight:
984.25
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing