Skip to Content
Merck
Anti-TNFRSF14 antibody produced in rabbit affinity isolated antibody

Anti-TNFRSF14 antibody produced in rabbit

Anti-TNFRSF14 antibody produced in rabbit affinity isolated antibody
Synonym(s):
Anti-Herpesvirus entry mediator A, Anti-TR2, Anti-Tumor necrosis factor receptor superfamily member 14 precursor, Anti-Tumor necrosis factor receptor-like 2

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique