![Anti-PRKCA antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/145/653/247d04b0-d25a-46ed-9b9b-38829b0bc134/640/247d04b0-d25a-46ed-9b9b-38829b0bc134.jpg)
Anti-PRKCA antibody produced in rabbit
![Anti-PRKCA antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/145/653/247d04b0-d25a-46ed-9b9b-38829b0bc134/640/247d04b0-d25a-46ed-9b9b-38829b0bc134.jpg)
Synonym(s):
KPCA, PKC III, PKC-α, PKC-A, kinase PKC-α
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing