Skip to Content
Merck
Anti-LAMP1 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution

Anti-LAMP1 antibody produced in rabbit

Anti-LAMP1 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-CD107a, Anti-LAMPA, Anti-LGP120, Anti-Lysosomal-associated membrane protein 1, Anti-Lysosome associated membrane protein 1, Anti-CD107a antigen, Anti-LAMP-1, Anti-Lysosome-associated membrane glycoprotein 1 precursor

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique