Skip to Content
Merck
Anti-IRF4 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-IRF4 antibody produced in rabbit

Anti-IRF4 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-IRF-4 antibody produced in rabbit, Anti-Interferon regulatory factor 4 antibody produced in rabbit, Anti-LSIRF antibody produced in rabbit, Anti-Lymphocyte-specific interferon regulatory factor antibody produced in rabbit, Anti-Multiple myeloma oncogene 1 antibody produced in rabbit, Anti-NF-EM5 antibody produced in rabbit, IRF3, Interferon regulatory factor 3

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique