Anti-HDAC2 antibody produced in rabbit
Synonym(s):
Anti-HD2, Anti-Histone deacetylase 2
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3