Anti-DNAJB11 antibody produced in rabbit
Synonym(s):
Anti-DnaJ homolog subfamily B member 11 precursor antibody produced in rabbit, Anti-ER-associated Hsp40 co-chaperone antibody produced in rabbit, Anti-ER-associated dnaJ protein 3 antibody produced in rabbit, Anti-ErJ3 antibody produced in rabbit, Anti-PWP1-interacting protein 4 antibody produced in rabbit, Anti-hDj9 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing