![Anti-DAZAP1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/257/314/341b2529-9791-4a36-a3b2-9b59f0e34623/640/341b2529-9791-4a36-a3b2-9b59f0e34623.jpg)
Anti-DAZAP1 antibody produced in rabbit
![Anti-DAZAP1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/257/314/341b2529-9791-4a36-a3b2-9b59f0e34623/640/341b2529-9791-4a36-a3b2-9b59f0e34623.jpg)
Synonym(s):
Anti-DAZ-associated protein 1 antibody produced in rabbit, Anti-Deleted in azoospermia-associated protein 1 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing