Anti-BANF1 antibody produced in rabbit
Synonym(s):
Anti-BAF
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing