![Anti-ALDH9A1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/303/183/c4cf1a69-2358-4085-8d98-7fdb02f9ba7d/640/c4cf1a69-2358-4085-8d98-7fdb02f9ba7d.jpg)
Anti-ALDH9A1 antibody produced in rabbit
![Anti-ALDH9A1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/303/183/c4cf1a69-2358-4085-8d98-7fdb02f9ba7d/640/c4cf1a69-2358-4085-8d98-7fdb02f9ba7d.jpg)
Synonym(s):
Anti-γ-Aminobutyraldehyde dehydrogenase antibody produced in rabbit, Anti-4-Trimethylaminobutyraldehyde dehydrogenase antibody produced in rabbit, Anti-Aldehyde dehydrogenase 9A1 antibody produced in rabbit, Anti-Aldehyde dehydrogenase E3 isozyme antibody produced in rabbit, Anti-R-aminobutyraldehyde dehydrogenase antibody produced in rabbit, Anti-TMABADH antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1