Skip to Content
Merck
Acetic anhydride-d6 99 atom % D

Acetic anhydride-d6

Acetic anhydride-d6 99 atom % D
Linear Formula:
(CD3CO)2O
CAS Number:
Molecular Weight:
108.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing