![4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/150/215/ff680071-f4af-46db-b69a-d343b6b52e03/640/ff680071-f4af-46db-b69a-d343b6b52e03.png)
4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone
![4-(Methylnitrosoamino)-1-(3-pyridinyl)-1-butanone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/150/215/ff680071-f4af-46db-b69a-d343b6b52e03/640/ff680071-f4af-46db-b69a-d343b6b52e03.png)
Synonym(s):
4-(Methylnitrosamino)-1-(3-pyridinyl)-1-butanone, NNK
Empirical Formula (Hill Notation):
C10H13N3O2
CAS Number:
Molecular Weight:
207.23
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing