Skip to Content
Merck
4-Hydroxy-3-methoxyphenylglycol hemipiperazinium salt

4-Hydroxy-3-methoxyphenylglycol hemipiperazinium salt

4-Hydroxy-3-methoxyphenylglycol hemipiperazinium salt
Synonym(s):
3-Methoxy-4-hydroxy-phenylglycol piperazine, MHPG piperazine, MOPEG, MOPEG piperazine
Empirical Formula (Hill Notation):
C9H12O4 · 0.5C4H10N2
CAS Number:
Molecular Weight:
227.26
EC Number:

Products

Product Category
Brand
Formula Weight
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing