
3-Hydroxy-2-methylbutyric acid, mixture of isomers

Synonym(s):
3-Hydroxy-2-methylbutanoic acid, Nilic acid
Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13
Products
Product Category
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing