Skip to Content
Merck
2-Ethyl-3(5 or 6)-dimethylpyrazine, mixture of isomers ≥95%, FG

2-Ethyl-3(5 or 6)-dimethylpyrazine, mixture of isomers

2-Ethyl-3(5 or 6)-dimethylpyrazine, mixture of isomers ≥95%, FG
Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing