
2-Chloro-4-iodo-nicotinic acid methyl ester

Empirical Formula (Hill Notation):
C7H5ClINO2
CAS Number:
Molecular Weight:
297.48
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing