Skip to Content
Merck
2-Bromoterephthalic acid 95%

2-Bromoterephthalic acid

2-Bromoterephthalic acid 95%
Linear Formula:
BrC6H3-1,4-(CO2H)2
CAS Number:
Molecular Weight:
245.03
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing