Skip to Content
Merck
2-Acetyl-3-methylpyrazine ≥98%, FG

2-Acetyl-3-methylpyrazine

2-Acetyl-3-methylpyrazine ≥98%, FG
Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing