Skip to Content
Merck
2,5-Dichlorobenzotrifluoride 98%

2,5-Dichlorobenzotrifluoride

2,5-Dichlorobenzotrifluoride 98%
Synonym(s):
2,5-Dichloro-α,α,α-trifluorotoluene
Linear Formula:
Cl2C6H3CF3
CAS Number:
Molecular Weight:
215.00
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing