1-Naphthalenyl ester fluorosulfuric acid
Empirical Formula (Hill Notation):
C10H7FO3S
CAS Number:
Molecular Weight:
226.22
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing