
11-Ketofluticasone Propionate

Synonym(s):
6alpha, 9-difluoro-17- [[(fluoromethyl)sulfanyl] carbonyl]-16alpha-methyl-3,11-dioxoandrosta-1-4dien-17alpha-yl-propanoate
Empirical Formula (Hill Notation):
C25H29F3O5S
CAS Number:
Molecular Weight:
498.55
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing