Skip to Content
Merck

SY3H9888F4FC

Synthonix Corporation

tert-Butyl 2-(oxetan-3-ylidene)acetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
170.21
MDL number:

Pricing and availability is not currently available.

Iupac Name

tert-butyl 3-oxetanylideneacetate

Inchi Code

1S/C9H14O3/c1-9(2,3)12-8(10)4-7-5-11-6-7/h4H,5-6H2,1-3H3

InChI key

ARJYDTSMXYEQSN-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
8.51009128058.51087
BOP Benzotriazole-1-yl-oxy-tris-(dimethylamino)-phosphonium hexafluorophosphate Novabiochem®

8.51004

BOP

PyBOP® Novabiochem®

8.51009

PyBOP®

PyClock 6-Chloro-benzotriazole-1-yloxy-tris-pyrrolidinophosphonium hexafluorophosphate Novabiochem®

8.51087

PyClock

assay

≥99.0% (HPLC)

assay

≥99.0% (HPLC)

assay

≥97.0% (TLC)

assay

≥98.0% (HPLC)

Quality Level

200

Quality Level

400

Quality Level

200

Quality Level

200

manufacturer/tradename

Novabiochem®

manufacturer/tradename

Novabiochem®

manufacturer/tradename

-

manufacturer/tradename

Novabiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

product line

Novabiochem®

product line

Novabiochem®

product line

-

product line

Novabiochem®

storage temp.

Refrigerator

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

liquid

Product Link

https://synthonix.com/7166.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://synthonix.com/7166.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service