Skip to Content
Merck
All Photos(1)

Key Documents

EHU057281

Sigma-Aldrich

MISSION® esiRNA

targeting human E2F3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACCCTGGACCTCAAACTGTTAACCGAGGATTCAGAGAATCAAAGGTTAGCTTATGTTACATATCAAGATATTCGAAAAATTAGTGGCCTTAAAGACCAAACTGTTATAGTTGTGAAAGCCCCTCCAGAAACAAGACTTGAAGTGCCTGACTCAATAGAGAGCCTACAAATACATTTGGCAAGTACCCAAGGGCCCATTGAGGTTTACTTATGTCCAGAAGAGACTGAAACACACAGTCCAATGAAAACAAACAACCAAGACCACAATGGGAATATCCCTAAACCCGCTTCCAAAGACTTGGCTTCAACCAACTCAGGACATAGCGATTGCTCAGTTTCTATGGGAAACCTTTCTCCTCTGGCCTCCCCAGCCAACCTCTTACAGCAGACTGAGGACCAAATTCCTTCCAACCTAGAAGGACCGTTTGTGAACTTACTGCCTCCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoko Nakashima et al.
European journal of dermatology : EJD, 27(5), 464-471 (2017-07-26)
Although angiosarcoma exhibits aggressive progression and is associated with unfavourable prognosis, its pathogenesis is poorly understood. In the present study, we investigated the possibility that microRNAs play a role in the pathogenesis of angiosarcoma. microRNA expression was evaluated by array
Junsheng Guo et al.
International journal of clinical and experimental pathology, 13(3), 587-596 (2020-04-10)
Bladder cancer is a common, serious disease worldwide. MicroRNAs (miRNAs) have been reported to participate in the development and progression in many cancers, including bladder cancer. However, the exact roles of miR-22 in bladder cancer process and its underlying mechanism
Zhicai Feng et al.
OncoTargets and therapy, 11, 5303-5313 (2018-09-15)
Melanoma is a malignant tumor that seriously affects patients. The pathogenesis of malignant melanoma is complex, and the cell cycle is closely related to tumor progression. Based on the catalog of cancer somatic mutations, we found that overexpression of the
Sivakumar Vadivel Gnanasundram et al.
Nature communications, 8(1), 2103-2103 (2017-12-14)
The c-myc oncogene stimulates ribosomal biogenesis and protein synthesis to promote cellular growth. However, the pathway by which cells sense and restore dysfunctional mRNA translation and how this is linked to cell proliferation and growth is not known. We here
Dianzhong Geng et al.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society, 25(4), 707-713 (2015-02-13)
Previous studies confirmed that high-risk human papillomavirus (HR-HPV) infection is a risk factor of cervical cancer, and the infection was associated with significantly reduced miR-34a expression during carcinogenesis. However, the downstream targets of miR-34a and their roles are still not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service