Skip to Content
Merck
All Photos(1)

Key Documents

8.26001

Sigma-Aldrich

2-(Acetylaminomethyl)-chromen-4-one

≥97% (HPLC)

Synonym(s):

2-(Acetylaminomethyl)-chromen-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
UNSPSC Code:
12352111
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

solid

storage temp.

2-30°C

InChI

1S/C12H11NO3/c1-8(14)13-7-9-6-11(15)10-4-2-3-5-12(10)16-9/h2-6H,7H2,1H3,(H,13,14)

InChI key

YPANBNNTDTUMLB-UHFFFAOYSA-N

General description

for synthesis

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Water (K. F.): ≤ 5.0 %
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service