Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249D213

J & W Pharmlab LLC

1H-Pyrazolo[3,4-c]pyridin-7-ylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
170.6
MDL number:

Iupac Name

1H-pyrazolo[3,4-c]pyridin-7-amine hydrochloride

Inchi Code

1S/C6H6N4.ClH/c7-6-5-4(1-2-8-6)3-9-10-5;/h1-3H,(H2,7,8)(H,9,10);1H

InChI key

QNWSABAYVBUIFD-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Light Yellow Solid

Product Link

https://www.jwpharmlab.com/home/product/45R0549S

MSDS

https://www.jwpharmlab.com/home/product/45R0549S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service