Merck
  • Noradrenaline, oxymetazoline and phorbol myristate acetate induce distinct functional actions and phosphorylation patterns of α
Noradrenaline, oxymetazoline and phorbol myristate acetate induce distinct functional actions and phosphorylation patterns of α
Biochimica et biophysica acta (2017-09-11)
Rocío Alcántara-Hernández, Aurelio Hernández-Méndez, M Teresa Romero-Ávila, Marco A Alfonzo-Méndez, André S Pupo, J Adolfo García-Sáinz
ABSTRACT
In LNCaP cells that stably express α
Sigma-Aldrich
5-Methylurapidil, solid