Merck
  • Reduced BAFF-R and increased TACI expression in common variable immunodeficiency.
Reduced BAFF-R and increased TACI expression in common variable immunodeficiency.
Journal of clinical immunology (2014-05-09)
Rita R Barbosa, Susana L Silva, Sara P Silva, Alcinda C Melo, M Conceição Pereira-Santos, João T Barata, Lennart Hammarström, Marília Cascalho, Ana E Sousa
ABSTRACT
B-cell survival and differentiation critically depend on the interaction of BAFF-R and TACI with their ligands, BAFF and APRIL. Mature B-cell defects lead to Common Variable Immunodeficiency (CVID), which is associated with elevated serum levels of BAFF and APRIL. Nevertheless, BAFF-R and TACI expression in CVID and their relationship with ligand availability remain poorly understood. We found that BAFF-R expression was dramatically reduced on B cells of CVID patients, relative to controls. BAFF-R levels inversely correlated with serum BAFF concentration both in CVID and healthy subjects. We also found that recombinant BAFF stimulation reduced BAFF-R expression on B cells without decreasing transcript levels. On the other hand, CVID subjects had increased TACI expression on B cells in direct association with serum BAFF but not APRIL levels. Moreover, splenomegaly was associated with higher TACI expression, suggesting that perturbations of TACI function may underlie lymphoproliferation in CVID. Our results indicate that availability of BAFF determines BAFF-R and TACI expression on B cells, and that BAFF-R expression is controlled by BAFF binding. Identification of the factors governing BAFF-R and TACI is crucial to understanding CVID pathogenesis, and B-cell biology in general, as well as to explore their potential as therapeutic targets.
Sigma-Aldrich
BAFF active human, Animal-component free, recombinant, expressed in Nicotiana, >97% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
BAFF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Pricing and availability is not currently available.