Merck
  • Effect of temperature and atmospheric environment on the photodegradation of some Disperse Red 1-type polymers.
Effect of temperature and atmospheric environment on the photodegradation of some Disperse Red 1-type polymers.
Optics letters (2007-12-15)
A Galvan-Gonzalez, M Canva, G I Stegeman, R Twieg, T C Kowalczyk, H S Lackritz
ABSTRACT
The photodegradation of the azobenzene chromophore DR1 {4-[N-ethyl-N-(2-hydroxyethyl)amino]-4(?)-nitroazobenzene]} incorporated as a side chain or as a guest in a poly(methyl) methacrylate host has been evaluated as a function of wavelength, temperature, and the atmospheric environment. The effects of these variables on the lifetime of DR1-based electro-optic devices is quantified.
Sigma-Aldrich
Disperse Red 19, Dye content 97 %