Merck
  • Tumour initiating activity of dihydrodiols of benzo[b]fluoranthene, benzo[j]fluoranthene, and benzo[k]fluoranthene.
Tumour initiating activity of dihydrodiols of benzo[b]fluoranthene, benzo[j]fluoranthene, and benzo[k]fluoranthene.
Carcinogenesis (1982-01-01)
E J LaVoie, S Amin, S S Hecht, K Furuya, D Hoffmann
ABSTRACT
The tumor initiating activities on mouse skin of benzo[b]fluoranthene, (B[b]F), benzo[j]fluoranthene (B[j]F), benzo[k]fluoranthene (B[k]F] and three of their dihydrodiols, 9,10-dihydro-9,10-dihydroxybenzo[b]fluoranthene (B[b]F-9,10-diol), 9,10-dihydro-9,10-dihydroxybenzo[j]fluoranthene (B[j]F-9,10-diol), and 8,9-dihydro-8,9-dihydroxybenzo[k]fluoranthene (B[k]F-8,9-diol) were evaluated. Among the parent hydrocarbons, B[b]F was the most potent tumor initiator, with activity greater than that of B[j]F but less than that of benzo[a]pyrene. B[k]F also showed tumor initiating activity, in contrast to its lack of complete carcinogenic activity on mouse skin. B[b]F-9,10-diol, which can form a bay region dihydrodiol epoxide, was as active as B[b]F. B[j]F-9,10-diol, which would form its dihydrodiol epoxide in a four sided pseudo-bay region, was less active than B[j]F. B[k]F-8,9-diol was inactive. These results, together with parallel metabolic studies, suggest that the formation of bay region dihydrodiol epoxides may not be the major activation mechanism in benzofluoranthene tumorigenesis.
Product Description
Supelco
Benzo[j]fluoranthene solution, certified reference material, 2000 μg/mL in dichloromethane