Merck
  • Nyctiphanes couchii as intermediate host for Rhadinorhynchus sp. (Acanthocephala, Echinorhynchidae) from NW Iberian Peninsula waters.
Nyctiphanes couchii as intermediate host for Rhadinorhynchus sp. (Acanthocephala, Echinorhynchidae) from NW Iberian Peninsula waters.
Diseases of aquatic organisms (2013-07-10)
M Gregori, F J Aznar, E Abollo, A Roura, A F González, S Pascual
ABSTRACT
In the mesozooplanktonic community of the coastal upwelling system of the Ría de Vigo (NW Spain), the euphausiid Nyctiphanes couchii has been identified for the first time in temperate waters of the NE Atlantic as the intermediate host for cystacanths of Rhadinorhynchus sp. Parasites were identified using morphological characters described in 20 cystacanths. The hooks of the proboscis were arranged in 14 rows of 26 hooks each, while the hooks of the basal circle were only slightly erected and were longer than remaining spines. A maximum-likelihood estimation (ML) tree inferred from the 18S rRNA data set of Palaeacantocephala revealed that our specimens belong to a highly supported clade with Rhadinorhynchus sp., Pararhadinorhynchus sp. and Transvena annulospinosa. Nonetheless, our morphological and phylogenetic analyses suggested that the status of Rhadinorhynchus pristis should be re-examined. The prevalences of parasites were 0.0019% and 0.0001% for frontal and coastal summer communities, and 0.0068% and 0.0008% for coastal and oceanic autumn communities, respectively. The presence of these cystacanths in different mesozooplankton communities throughout the study suggests that the recruitment of parasites may be affected by the oceanography.
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization