Merck
  • Prospective multicenter clinical trial of Chinese herbal formula JZQG (Jiangzhuoqinggan) for hypertension.
Prospective multicenter clinical trial of Chinese herbal formula JZQG (Jiangzhuoqinggan) for hypertension.
The American journal of Chinese medicine (2013-01-23)
Xiao-Lin Tong, Feng-Mei Lian, Qiang Zhou, Li-Peng Xu, Hang-Yu Ji, Gui-Cheng Xu, Yuan-Hui Hu, Lin-Hua Zhao, Le Xia, Jia Wang, Xin-Yan Chen, Man-Hon Chan, Lan-Lan Zhang, Wen Gao, Zhong Zhen, Shui-Ping Zhou, Bai Chang
ABSTRACT
A prospective multicenter clinical trial was conducted to compare the beneficial effects of a Chinese herbal medicine formula Jiangzhuoqinggan (JZQG) and western antihypertension drug irbesartan. JZQG is mainly composed of rhubarb, coptis, cassia, and uncaria. A total of 240 patients with mild to moderate hypertension were enrolled in the trial. Patients were assigned into two groups after screening: JZQG group and the irbesartan group. After four weeks of treatment, we compared the changes in routine blood pressure, 24 h ambulatory blood pressure, and waist circumference. There was a significant reduction in systolic blood pressure and diastolic blood pressure in the JZQG group (both p < 0.01). There were no significant differences between the reduction of systolic and diastolic blood pressures in the two treatment groups. From the 24 h ambulatory blood pressure measurement, the JZQG group showed a greater reduction in both systolic and diastolic blood pressures (in both daytime and nighttime) than the irbesartan group. Furthermore, there was a significant difference in waist circumference in the JZQG group (1.51 cm reduction; P < 0.05) but not the irbesartan group (0.42 cm). Thus, the JZQG formula may have therapeutic value in patients with both hypertension and metabolic syndrome.
Sigma-Aldrich
Irbesartan, ≥98% (HPLC), powder
Irbesartan, European Pharmacopoeia (EP) Reference Standard